Biox Culture Dishes
Supplier: TWD Scientific
BX07-3035 BX07-3060 BX07-3100 BX07-3150
76081-548CS 439.81 CAD
76081-548 76081-550 76081-552 76081-554
Biox Culture Dishes
Culture Dishes Cell Culture Dishes
Sterile and constructed of optically clear material, these cell culture dishes offer high quality at a great price.
  • High-quality medical grade polystyrene
  • Vacuum plasma surface treatment ensures excellent cell adhesion
  • Lid design promotes effective gas exchange
  • External grip for easier handling (available on 2 models)
  • Electron beam sterilized
  • Non-pyrogenic and non-cytotoxic
  • Dnase- & Rnase-free, human DNA-free
These TC treated culture dishes have vented lids and raised edges for convenient stacking.
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008.
Order Now
SPECIFICATIONS
Environmentally Preferable