LB Agar Plates, Teknova
Supplier: TEKNOVA CA
L5920
L1920
L1975
CA100217-690PK
1170.53
CAD
CA100217-690
CA100219-336
CA100219-348
LB Agar Plates, Teknova
Media for Microbiology
Prepared Media for Microbiology
LB is a nutritionally rich growth medium used for the cultivation of recombinant strains of Escherichia coli and enteric bacteria
- Shipping conditions: Ambient
- Storage conditions: Refrigerate
These sterile pre poured plates are enriched with tryptone, yeast extract, sodium chloride, agar, X-gal and IPTG.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...