Biox Tissue Culture Flasks
Supplier: TWD Scientific
07-8025 BX07-9025 BX07-9075
76081-540CS 495.44 CAD
76081-540 76081-542 76081-544 76085-626
Biox Tissue Culture Flasks
Flasks Cell Culture Flasks
High quality polystyrene cell culture flasks are sterile, stackable, and a great value.
  • High-quality medical grade polystyrene
  • Plug caps completely isolate the culture from the outside environment
  • Filter Caps prevent cross-contamination while meeting the requirements for gas exchange
  • Electron beam sterilized
  • Short, wide, angled neck design for easy access
  • Non-pyrogenic and non-cytotoxic
  • DNase- & RNase-free, Human DNA-free
TC treated cell culture flasks with plug or filter caps allow for complete isolation from the outside environment and prevent cross contamination while allowing for gas exchange.
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008.