Magne-Box™ Magnetic Polycarbonate Cryoboxes, Argos Technologies
Supplier: Antyila Scientific
04200-03 04200-04 04200-05 04200-06
76102-168PK 552.48 CAD
76102-168 76102-170 76102-172 76102-174
Magne-Box™ Magnetic Polycarbonate Cryoboxes, Argos Technologies
Boxes Cryoboxes
Magne-Box™ freezer boxes are the first of its kind to offer a magnetic lid closure.
  • Boxes fit in standard size freezer racks
  • Polycarbonate design withstands temperatures from –196 to 121 °C
  • Slot openings on the bottom of boxes allow for drainage
  • 25-, 81-, and 100-Place boxes accommodate 2 mL cryovials
  • 81-Place Tall boxes accommodate 4 to 5 mL cryovials
Each box features magnets that securely connect the base to the hinged lid. This allows one-handed retrieval from freezer racks and ensures that samples stay safe. Polycarbonate boxes have clear lidsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable