Pig Recombinant IL-8 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
400-03-5UG 400-03-20UG 400-03-100UG 400-03-500UG 400-03-1MG
75845-248EA 133.4 CAD
75845-248 75845-250 75845-252 75845-254 75845-308
Pig Recombinant IL-8 (from E. coli)
Proteins and Peptides
Interleukin 8 (IL-8 or CXCL8) is a member of the CXC cytokine family and is produced by macrophages, epithelial, smooth muscle, and endothelial cells. IL-8 binds the  G protein-coupled serpentine receptors CXCR1 and CXCR2. IL-8 recruits innate immune cells, induces phagocytosis, and stimulates angiogenesis.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.