Mouse Recombinant MIP-2 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Insulin-like growth factor II (IGF-II) is an important fetal growth hormone made by theca cells during gestation. IGF-II engages the IGF-I receptor (IGF1R) to mediate embryonic growth. IGF-II also binds the sink IGF-II receptor (IGF2R) leading to IGF-II degradation.
- High quality
- Low endotoxin
- Affordable
Macrophage Inflammatory Protein 2 (MIP-2), also known as CXCL2, is a small cytokine that is secreted by monocytes and neutrophils at sites of inflammation. MIP-2 functions through the chemokine receptor CXCR2 to act as a chemotactic agent for leukocytes and hematopoietic cells.
Caution: For research use only.