Mouse Recombinant MIF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-78-5UG
200-78-25UG
200-78-100UG
200-78-1MG
200-78-500UG
10821-800EA
136.99
CAD
10821-800
10821-802
10821-804
10821-806
10822-278
Mouse Recombinant MIF (from E. coli)
Proteins and Peptides
Macrophage migration inhibitory factor (MIF) is a pro-inflammatory lymphokine that functions during cell-mediated immmunity. MIF promotes fibroblast migration by inducing interleukin 1 (IL-1), interleukin 8 (IL-8), and matrix metalloproteinase (MMP) expression. In interferon-gamma-activated macrophages, MIF stimulates nitric oxide (NO) production and tumor necrosis factor alpha (TNF-α) secretion.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.