Mouse Recombinant IL-22 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-77-10UG 200-77-100UG 200-77-1MG 200-77-500UG
10821-600EA 143.21 CAD
10821-600 10821-602 10821-604 10821-606 10822-204
Mouse Recombinant IL-22 (from E. coli)
Proteins and Peptides
Interleukin 22 (IL-22), also called IL-TIF, is an IL-10 family member that is produced by activated dendritic cells and T lymphocytes. IL-22 signals via a heteroduplex receptor consisting of IL-22R and IL-10RB chains. IL-22 is a potent mediator of cellular inflammatory responses and wound healing.  
  • High Quality
  • Low Endotoxin
  • Affordable
Caution: For research use only.