Mouse Recombinant IL-19 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Interleukin-19 (IL-19) is a member of the interleukin 10 (IL-10) cytokine family and is produced by B cells and monocytes. IL-19 binds the interleukin 20 receptor complex (IL-20R) to activate STAT3 signaling. IL-19 induces interleukin 6 (IL-6) and tumor necrosis factor alpha (TNF-α) expression in monocytes, and promotes type 2 T helper (Th2) cell-mediated immune responses. IL-19 production is upregulated in resting monocytes following granulocyte-macrophage colony-stimulating factor (GM-CSF) or lipopolysaccharide (LPS) stimulation.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.