Human Recombinant ENA 78 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-185-5UG 100-185-20UG 100-185-100UG 100-185-1MG 100-185-500UG
10821-640EA 136.99 CAD
10821-640 10821-642 10821-644 10821-646 10821-932
Human Recombinant ENA 78 (from E. coli)
Proteins and Peptides
Epithelial-derived neutrophil-activating peptide 78 (ENA 78), also known as CXCL5, is a chemokine that recruits neutrophils, promotes angiogenesis, and stimulates connective tissue remodelling. ENA 78 production is stimulated by interleukin 1 (IL-1) or tumor necrosis factor alpha (TNFα), and signals through the chemokine receptor CXCR2. ENA 78, 5-78aa is one of three naturally occuring ENA 78 variants in which the N-terminus has been truncated.
  • High Quality
  • Low Endotoxin
  • Affordable
Caution: For research use only.