Human Recombinant Exodus-2 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-06-20UG 100-06-100UG 100-06-500UG 100-06-5UG 100-06-1MG
75845-236EA 333.17 CAD
75845-236 75845-238 75845-260 75845-262 76051-284 77660-654
Human Recombinant Exodus-2 (from E. coli)
Proteins and Peptides
Exodus-2, also known as CCL21 and 6Ckine, is a chemokine that is strongly produced in the human lymph nodes and spleen. Exodus-2 signals through the chemokine receptor CCR7 to regulate thymocyte and activated T cell migration. Exodus-2 also mediates the homing of lymphocytes to the lymphatic system.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.