Ultrasonic Test Sieve Cleaner, WS Tyler Inc
Supplier: W.S. Tyler
P-1405
57332-230EA 7078.32 CAD
57332-230
Ultrasonic Test Sieve Cleaner, WS Tyler Inc
Ultrasonic Cleaners
Ultrasonic test sieve cleaner produces ultrasonic vibrations in a cleaning fluid bath, which causes contaminants to become dislodged from both sieve frame and media.
  • Compact and portable
  • Stainless steel oscillation tank
  • Built-in timer
Ordering information: Solution (mild detergent recommended) has to be ordered separately.