LB Agar Plate with Tetracycline, Teknova
Supplier: TEKNOVA CA
L1032
L2032
L1981
L1067
CA100216-602PK
833.07
CAD
CA100216-602
CA100217-268
CA100218-454
CA100220-114
CA101319-934
LB Agar Plate with Tetracycline, Teknova
Media for Microbiology
Prepared Media for Microbiology
These are sterile pre poured LB agar plates enriched with tryptone, yeast extract, sodium chloride, agar and tetracycline
- Bacteriostatic
- Inhibits bacterial synthesis by binding to 30S ribosome
- Stock solutions are stable for 4 days
- Store at −20 °C
Tetracycline is light-sensitive and is degraded by exposure to UV light. It is used as genetic marker in mammalian cell culture since no 30S ribosome.