Iodine D'Antoni solution, dark brown liquid

Supplier: Ricca Chemical

4103-16 4103-32 4103-4
RC4103-16EA 230.68 CAD
RC4103-16 RC4103-32 RC4103-4
Iodine D'Antoni solution, dark brown liquid
Iodine D'Antoni

D’Antoni’s Iodine stain is recommended for use in wet mount preparations for detection of protozoan cysts and helminth eggs and larvae.


Color: Dark brown liquid
REAGENTS (CLASSICAL FORMULA)*
Iodine (CAS 7553-56-2)...........................................................15.0 g
Potassium Iodide (CAS 7681-11-0).........................................10.0 g
Demineralized Water (CAS 7732-18-5) ...............................1000.0 ml
*Adjusted as required to meet performance standards.

UN: 3264
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR