Caps for TrayCell™, PerkinElmer
Supplier: PerkinElmer
L7110292 L7110293
CA97024-714EA 723.85 CAD
CA97024-714 CA97024-716 CA97024-718 CA97024-720
Caps for TrayCell™, PerkinElmer
Closures Test/Sample Tube Closures
The TrayCell™ caps are fiber-optic ultra-micro measuring cells designed for the UV/Vis analysis of DNA/RNA and proteins.
  • Suitable for almost all spectrophotometers
  • Ideally suited for very small measurement volumes: 0.7 to 10 L
  • Trouble-free measurement of the sample at different optical path lengths
  • Fast and simple cleaning of the optics before measuring the next sample
  • Excellent reproducibility and samples can be reused after the measurement simply by piping them off
The dimensions of the TrayCell™ are equivalent to a standard cuvette in order to work in most spectrophotometers. Caps with dilution factors: 5, 10, 50 and 100.