Luna® Universal One-Step RT-qPCR Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E3005L E3005S E3005X E3005E E3005E
CA103307-248EA 943 CAD
CA103307-248 CA103307-250 CA103501-458 CA76169-440 76196-380
Luna® Universal One-Step RT-qPCR Kit, New England Biolabs
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Rapid, sensitive and precise dye-based qPCR detection and quantitation of RNA targets.
In the Luna One-Step RT-qPCR Kit, Hot Start Taq DNA Polymerase is combined with a novel WarmStart-activated reverse transcriptase, allowing dual control of enzyme activity via reversible, aptamer-based inhibition.
The Luna Universal One-Step Reaction Mix is supplied at 2X concentration and contains Hot-Start Taq DNA Polymerase, dNTPs, a fluorescent dsDNA-binding dye, and all required buffer components. It is formulated with a unique passive reference dye that is compatible across a variety of instrument platforms, including those that require a high or low ROX reference signal. The Reaction Mix also features dUTP for carryover prevention and a non-fluorescent visible dye for monitoring reaction setup. This visible dye does not overlap spectrally with the included dsDNA-binding dye and does not interfere with real-time detection.
The Luna WarmStart RT Enzyme Mix is supplied at 20X concentration and contains Luna WarmStart Reverse Transcriptase as well as Murine RNase Inhibitor to aid in preventing RNA degradation . It is compatible with various RNA sample types (total RNA, poly(A)-RNA, etc.) and sources.
The Luna WarmStart RT Enzyme Mix is supplied at 20X concentration and contains Luna WarmStart Reverse Transcriptase as well as Murine RNase Inhibitor to aid in preventing RNA degradation. It is compatible with various RNA sample types (total RNA, poly(A)-RNA, etc.) and sources.
This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. The engineered WarmStart Luna Reverse Transcriptase also possesses higher thermostability than many other RTs, allowing an optimal reaction temperature of 55 °C. For difficult targets/templates, higher RT step temperatures of up to 60 °C can be used without compromising Luna performance.