Sample Wrapper, Thomas Scientific
Supplier: Thomas Scientific
6514F70
48956-142PK 275.27 CAD
48956-142
Sample Wrapper, Thomas Scientific
Seals General Purpose Sealing Films
Sample Wrapper absorbs heat quickly for infrared ignition; die-cut from unsized paper
Sample area 32 x 30 mm, with 8 x 38 mm extending strip as fuse for ignition.