Auto Series Plus Automatic Hydraulic Laboratory Presses, Carver
Supplier: Carver
3888.1PL0000-115V 3889.4PL0000-230V 3891.4PL0000-230V 3892.4PL0000-230V 3893.4PL0000-230V 3894.4PL0000-230V 3895.4PL0000-230V
53880-030EA 63675.07 CAD
53880-030 53880-032 53880-036 53880-040 53880-042 53880-044 53880-046
Auto Series Plus Automatic Hydraulic Laboratory Presses, Carver
Presses
Auto Series Plus Automatic Hydraulic Laboratory Presses feature a 7" color operator interface, enhanced platen temperature control with auto-tuning, ramp heating capability, real-time trending of force and temperature.
  • Storage of up to 250 recipes
  • Ramp heating capability and Recipe preview
  • Seven day timer for auto platen heat-up (optional)
  • Datalogging of cycle parameters (optional)
  • Access to logged data through Wi-Fi (optional)
  • 48 ton models include a wrap-around floor stand
Auto Series Plus press models are available with 15 to 48 tons for clampingMore Product Information