J-KEM® Temperature Controller, Model 210/T, With Timer, Chemglass
Supplier: Chemglass
CG-3202-01 CG-3202-02 CG-3202-03
80062-918EA 3216.19 CAD
80062-918 80062-920 80062-922
J-KEM® Temperature Controller, Model 210/T, With Timer, Chemglass
Controllers Temperature Controllers
This model is sufficient for heating mantles up to 5 L in size along with most laboratory ovens and hot plates
This model also features an additional 100 hour digital timer to turn heating ON or OFF when time has expired.
Controller has the ability to display temperature without heating. Control temperature to within ± 0.1°C of liquids from 1 µL to 5 L.
Delivery information: Includes 1/8” dia. x 12” long PTFE coated thermocouple, 10' coiled extension cord, and a 14/20 PTFE thermocouple adapter.