HiVeg™ Bennet’s Agar, HiMedia
Supplier: HiMedia Laboratories
MV694-2.5KG MV694-2.5KG MV694-500G MV694-5KG MV694-5KG MV694-500G
CA89229-052EA 7254.44 CAD
CA89229-052 89229-052 CA89229-050 CA61001-244 89229-050 61001-244
HiVeg™ Bennet’s Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Bennet’s HiVeg agar is used for cultivation and enhancement of sporulation of Nocardia and Streptomyces.
  • Light yellow coloured, may have slightly greenish tinge, homogeneous, free flowing powder
  • Storage below 30 °C