HiVeg™ Motility Indole Ornithine Medium, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
MV378-500G MV378-2.5KG MV378-5KG
CA61000-944EA 1141.5 CAD
CA61000-944 CA89228-574 CA89228-576
HiVeg™ Motility Indole Ornithine Medium, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Motility Indole Ornithine Medium is used for the identification of Enterobacteriaceae on the basis of motility, indole production and ornithine decarboxylase activity.
  • Light yellow coloured, may have slightly greenish tinge, homogeneous, free flowing powder
  • Storage below 30 °C