Funnels, Separatory, Squibb, PTFE Stopcock, With Sealed Drip Tip, Chemglass
Supplier: Chemglass
CG-1750-03 CG-1750-04 CG-1750-06 CG-1750-01 CG-1750-02
80071-150EA 504.79 CAD
80071-150 80071-152 80071-156 80071-146 80071-148
Funnels, Separatory, Squibb, PTFE Stopcock, With Sealed Drip Tip, Chemglass
Funnels Separating Funnels
Squibb-style separatory funnel with the plain lower stem replaced by an inner standard taper joint of the listed size having a sealed in drip tip.
Delivery information: Supplied complete with a flask length PTFE stopper.