Stirrer Shaft Couplings, Universal, Chemglass
Supplier: Chemglass
CG-2046-03 CG-2046-01 CG-2046-02 CG-2046-04 CG-2046-06 CG-2046-05
80062-272EA 398.98 CAD
80062-272 80062-268 80062-270 80062-274 89029-030 10170-288
Stirrer Shaft Couplings, Universal, Chemglass
Stirrers Stirring Shafts
Coupling incorporates a universal joint to compensate for misalignment of drive motor and glass stirrer shaft.
  • Nylon bushing compresses around shaft using an allen screw
  • Stainless steel stud attaches directly to the chuck of a stirrer motor
Ordering information: Supplied complete with an allen wrench. Please note that other than the CG-2097 Process Reactor Stirrer Shafts, Chemglass 19mm OD stirrer shafts have a reduced top diameter of 12.7mm. When ordering a universal coupling for a Chemglass 19mm OD stirrer shaft, use part number CG-2046-03.