Graduated Cylinder, Chemglass
Supplier: Chemglass
CG-1224-20 CG-1224-21
80060-650EA 83.89 CAD
80060-650 80060-652
Graduated Cylinder, Chemglass
Cylinders Measuring Cylinders
These large capacity cylinders are made of polypropylene
  • Has double scale and hex base for stability
  • Steam autoclavable at 121 °C
  • Safer and more convenient
The handle is generously sized for large hands but with a slender enough grip for smaller hands. Hand opening is 3 1/2” x 1 1/8
The handle on the Holdfast™ graduated cylinder makes pouring safer and more convenient.