Filtering Flask, Heavy Wall, Chemglass
Supplier: Chemglass
CG-1550-04 CG-1550-07 CG-1550-10 CG-1550-01
80070-148EA 354.69 CAD
80070-148 80070-154 80070-160 80070-142
Filtering Flask, Heavy Wall, Chemglass
Flasks Filter Flasks
These heavy wall Erlenmeyer shaped filtering flasks have a standard taper outer joint and a serrated hose connection for attachment to vacuum source.