WHEATON® Packer Bottles, PET, Wide Mouth, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W209690 W209691 W209692
10847-632CS 197.08 CAD
10847-632 10847-634 10847-636
WHEATON® Packer Bottles, PET, Wide Mouth, DWK Life Sciences
Bottles Plastic Bottles
Wide mouth plastic packer bottles are supplied with an attached screw cap.
  • Natural color
  • White polypropylene PE lined cap
  • PET provides both the clarity of the glass and safety of plastic
Wide mouth packer bottles are high premium quality.