Titan3™ PVDF Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
42213-PV 42204-PV 44504-PV 42225-PV 44513-PV
14232-574PK 676.29 CAD
14232-574 14234-968 14234-972 14235-038 14235-140
Titan3™ PVDF Syringe Filters, Thermo Scientific
Filters Syringe Filters
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
  • Superior membrane quality for HPLC
  • Low non-specific binding with exceptional chemical resistance
  • Hydrophilic membrane provides excellent flow rates and low binding coefficients (without pre-filter)
  • Certified for chromatographic performance
  • Compatible with a wide range of aqueous and organic-based sample environments
  • Recommended for HPLC and UHPLC
  • Able to filter solutions at
More Product Information