Titan3™ PTFE (Hydrophobic) Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
44504-NP 42225-NP 44513-NP 44525-NP 41025-NP 42204-NP 42225-PC
14232-522PK 271.49 CAD
14232-522 14232-746 14232-749 14232-752 14232-882 14234-952 14235-364
Titan3™ PTFE (Hydrophobic) Syringe Filters, Thermo Scientific
Filters Syringe Filters
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
  • Excellent chemical resistance for use with organic matrices
  • Naturally hydrophobic membrane provides excellent flow rates and high loading capacities
  • Exceptional temperature stability, able to filter solutions at temperatures up to 110 °C
  • Organic solvent recommended
Available in sizes and membrane types to meet any application need, Thermo Scientific™ Titan3™ Hydrophobic PTFE patented Syringe Filters provide cleaner sample extracts by removing interfering materials and fine particles.
Hydrophobic PTFE filter applications are aggressive organic, highly basic or hot solutions and transducer protection. Filter aqueous solutions after pre-wetting with an alcohol. Due to the membrane's hydrophobic nature, PTFE filters can be utilized as a moisture barrier in venting applications.
Low extractable membranes and housings ensure that no filter-borne contaminants are added to your sample. Performance-tested under actual HPLC conditions. Integral 1 mm borosilicate glass pre-filter (30 mm devices) ensures greater confidence with high solids samples. Reinforcing ring provides greater strength to the housing to prevent leakage and bursting (maximum pressure for 30 mm devices 15 bar / 210 psi). Housings are manufactured from solvent-resistant, low-extractable polypropylene resins specifically selected for wide compatibility with common HPLC sample matrices.
Color coding makes it easy to select the correct membrane and pore size. Packed in re-usable rigid transparent color-coded containers. Syringe filters can be sterilized by autoclave at 121 °C for 15 minutes. Enhanced Luer lock inlet prevents leakage; outlet fitting is a standard size male Luer-slip fitting for ease of filtrate collection.
Caution: These products are intended for research only and are not to be used for any other purpose, including without limitation, unauthorized commercial use, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses, or any type of consumption by or application to humans or animals.