Titan3™ PES Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
42225-PS 44525-PS 42213-PS 44513-PS
14235-042PK 412.39 CAD
14235-042 14235-044 14235-146 14235-148
Titan3™ PES Syringe Filters, Thermo Scientific
Filters Syringe Filters
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
  • Precise results in sensitive analysis of ionic analytes
  • Certified for low-level IC interference by ICP analysis
  • Hydrophilic membrane provides excellent flow rates and low binding coefficients
  • Low affinity for binding drugs, ideal for dissolution testing procedures
  • Able to filter solutions at temperatures up to 100 °C
Available in sizes and membrane types to meet anyMore Product Information