Titan3™ Nylon (Polyamide) Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
44525-NC 44513-NN 41225-NN 42225-NN 44525-NN 45025-NN 42204-NN 44504-NN 44526-NN 42213-NN
14232-370CS 2395.54 CAD
14232-370 14232-576 14232-740 14232-744 14232-750 14232-756 14234-950 14234-954 14235-032 14235-132
Titan3™ Nylon (Polyamide) Syringe Filters, Thermo Scientific
Filters Syringe Filters
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
  • Membrane of choice for analytical applications
  • Naturally hydrophilic with broad chemical resistance
  • Excellent flow rates and high-throughput loading
  • HPLC recommended
  • Able to filter solutions at temperatures up to 100 °C
Thermo Scientific™ Titan3™ Nylon (Polyamide) patented Syringe Filters provide cleaner sample extracts by removing interfering materials and fineMore Product Information