Titan3™ Glass Microfiber Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
40725-GM 42725-GM
14235-016PK 466 CAD
14235-016 14235-020
Titan3™ Glass Microfiber Syringe Filters, Thermo Scientific
Filters Syringe Filters
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
  • For large particulate removal
  • Increased sample throughput
  • Low extraction neutral borosilicate glass
  • Able to filter solutions at temperatures up to 110 °C
  • For use with viscous or particle-laden samples
Available in sizes and membrane types to meet any application need, Thermo Scientific™ Titan3™ GMF patented Syringe Filters provide cleaner sample extracts by removing interfering materials and fine particles.
Glass microfiber filter applications are clarification and pre-filtering of suspensions GMF filters are available in a range of porosities.
Low extractable membranes and housings ensure that no filter-borne contaminants are added to your sample. Reinforcing ring provides greater strength to the housing to prevent leakage and bursting (maximum pressure for 30 mm devices 15 bar / 210 psi). Housings are manufactured from solvent-resistant, low-extractable polypropylene resins specifically selected for wide compatibility with common HPLC sample matrices.
Color coding makes it easy to select the correct membrane and pore size. Packed in re-usable rigid transparent color-coded containers. Syringe filters can be sterilized by autoclave at 121 °C for 15 minutes. Enhanced Luer lock inlet prevents leakage; outlet fitting is a standard size male Luer-slip fitting for ease of filtrate collection.
Caution: These products are intended for research only and are not to be used for any other purpose, including without limitation, unauthorized commercial use, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses, or any type of consumption by or application to humans or animals.