Titan3™ Cellulose Acetate CA Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
42204-CA 44502-CA 42225-CA 44525-CA 42213-CA 44513-CA
14234-966PK 281.03 CAD
14234-966 14234-970 14235-024 14235-028 14235-130 14235-136
Titan3™ Cellulose Acetate CA Syringe Filters, Thermo Scientific
Filters Syringe Filters
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
  • For filtering of aqueous solutions or biological samples
  • Hydrophilic membrane provides excellent flow rates and low binding coefficients
  • Certified for chromatographic performance
  • Superior choice for biological assays, gel capsule dissolution testing, protein sample matrixes
  • Recommended for aqueous HPLC
  • Able to filter solutions at temperatures up to 110 °C
Available in sizes and membrane types to meet any application need, Thermo Scientific™ Titan3™ Cellulose Acetate patented Syringe Filters provide cleaner sample extracts by removing interfering materials and fine particles.
Cellulose Acetate syringe filters have a well defined pore structure, giving effective retention of particles without excessive pore blockage. Compatible with aqueous solutions. They are excellent filters for biological samples.
Low extractable membranes and housings ensure that no filter-borne contaminants are added to your sample. Performance-tested under actual HPLC conditions. Integral 1 mm borosilicate glass pre-filter (30 mm devices) ensures greater confidence with high solids samples. Reinforcing ring provides greater strength to the housing to prevent leakage and bursting (maximum pressure for 30 mm devices 15 bar/210 psi). Housings are manufactured from solvent-resistant, low-extractable polypropylene resins specifically selected for wide compatibility with common HPLC sample matrices.
Color coding makes it easy to select the correct membrane and pore size. Packed in re-usable rigid transparent color-coded containers. Syringe filters can be sterilized by autoclave at 121 °C for 15 minutes. Enhanced Luer lock inlet prevents leakage; outlet fitting is a standard size male Luer-slip fitting for ease of filtrate collection.
Caution: These products are intended for research only and are not to be used for any other purpose, including without limitation, unauthorized commercial use, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses, or any type of consumption by or application to humans or animals.