Aluminum Seals, Tear-Out, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
73820B-13 73820C-13 73820D-13 73820B-20 73820C-20 73820D-20 73820-20 73820-13
66004-024CS 357.31 CAD
66004-024 66004-026 66004-028 66004-030 66004-032 66004-034 66010-797 66010-795
Aluminum Seals, Tear-Out, Kimble Chase
Closures Chromatography Vial Closures
These tear-out seals are designed to secure septa and stoppers to bottles and vials.
  • Tamper-evident center tear-out feature protects target area
  • For use with 13 and 20 mm serum finished bottles and vials
  • Allow for easy access to septa
Order Now
SPECIFICATIONS
Environmentally Preferable