Christensen Citrate Agar, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M143-500G M143-2.5KG M143-5KG
CA61000-026EA 1158.54 CAD
CA61000-026 CA89224-028 CA89224-030
Christensen Citrate Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of enteric pathogens and coliforms on the basis of citrate utilization.
  • Light yellow to light pink homogeneous free flowing powder
  • Reaction of 2.48% w/v aqueous solution at 25 °C. pH : 6.9 ± 0.2
Store below 30 °C in tightly closed container and the prepared medium at 2 - 8 °C.