Kuderna-Danish Concentrator Tubes, Graduated, Chemglass
Supplier: Chemglass
CG-1316-02 CG-1316-11 CG-1316-12
80068-032EA 167 CAD
80068-032 80068-038 80068-040
Kuderna-Danish Concentrator Tubes, Graduated, Chemglass
Concentrators
These graduated concentrator tubes are mainly designed for use with Kuderna-Danish evaporative set-ups.
  • Can be stoppered for storage
  • Available with and without hooks