DPD Chlorine Reagent Powder Pop Dispenser, HF Scientific
Supplier: HF SCIENTIFIC INC
10504C 10506C
97058-872EA 939.34 CAD
97058-872 97058-874
DPD Chlorine Reagent Powder Pop Dispenser, HF Scientific
Environmental Test Kits
This portable dispenser accurately delivers the correct volume of DPD reagent for free or total chlorine
  • Powder dissolves quickly in sample, leaving no messy residue behind
  • No spill dispensing into most round or square cuvettes
  • Dispenser design prevents clogging
To use, simply invert the dispenser over sample and press the button to dispense reagent.
Delivery information: Supplied with enough DPD powder for 100 tests or in multi-packs providing 400 or 1000 tests.