Adapter, Distillation, Chemglass
Supplier: Chemglass
CG-1022-45 CG-1022-01 CG-1022-02 CG-1022-14 CG-1022-19
16002-602EA 630.58 CAD
16002-602 80065-816 80065-818 80065-820 80065-822
Adapter, Distillation, Chemglass
Distillation Components Distillation Adapters
These adapters for distillation systems are designed with standard taper outer joint at the top and inner joints on the bottom and sidearm.
  • Inner joints are at a 75° angle to one another
  • All the joints are in same size