Sample Vials, Amber, PTFE Lined Caps, Chemglass
Supplier: Chemglass
CG-4905-01 CG-4905-02 CG-4905-03 CG-4905-04 CG-4905-05 CG-4905-06
89029-458EA 785.33 CAD
89029-458 89029-460 89029-462 89029-464 89029-466 89029-468
Sample Vials, Amber, PTFE Lined Caps, Chemglass
Vials
Sample vials are made from Type 1 Borosilicate glass.
  • Full height partitions minimize breakage
  • Solid PTFE lined caps
Packaging: Packaged in convenient LAB-PACS of 144 pieces.
Order Now
SPECIFICATIONS
Environmentally Preferable