Fixable Viability Kit, Zombie Violet™
Supplier: Biolegend
Zombie Violet is an amine-reactive fluorescent dye that is non-permeant to live cells but permeant to the cells with compromised membranes
Zombie Violet™ dye is excited by the violet laser (405 nm) and has a fluorescence emission maximum of 423 nm. If using in a multi-color panel design, filter optimization may be required depending on other fluorophores used.
Thus, it can be used to assess live vs. dead status of mammalian cells. Zombie Violet is a polar, water soluble dye providing green fluorescence, making it suitable for multi-color detection.
Delivery information: Zombie Violet™ Fixable Viability Kit is composed of lyophilized Zombie Violet™ dye and anhydrous DMSO.