Scissors Lift Tables, Wesco
Supplier: Wesco
273261 273269 273230 273710 273711 273712 273713 260199 260200 260201 260204 260207 260208 272970 272973 272974 272862
76017-650EA 4646.02 CAD
76017-650 76017-654 76017-656 76017-658 76017-660 76017-662 76017-664 76017-874 76017-876 76017-878 76017-880 76017-882 76017-884 76017-886 76017-888 76017-890 76017-892
Scissors Lift Tables, Wesco
Laboratory Jacks
Scissors Lift tables are used to lift
  • Moldon polyurethane wheels and locking swivel casters
  • Capacities 330 - 2200 lbs
  • Chrome plated handle and foot pedal
  • Foot operated pump
  • Folding and fixed handles