Solvent Blue 38 ready-to-use

Supplier: POLYSCIENCES INC-RESEARCH CHEM

Synonyms: Direct Blue 86Luxol® Fast Blue MBSNC.I. 74180

24611-500
75805-994EA 419.71 CAD
75805-994
Solvent Blue 38 ready-to-use
Solvent Blue 38

Luxol fast blue, ready-to-use, is used to stain myelin and phospholipids.

Formula: C₃₂H₁₂CuN₈Na₂O₆S₂
MW: 778.15 g/mol
MDL Number: MFCD00071424
CAS Number: 1328-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR