Proteose Peptone A, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
RM6394-5KG RM6394-10KG RM6394-25KG RM6394-2.5KG RM6394-500G
CA95007-733EA 10406.28 CAD
CA95007-733 CA95007-735 CA95007-737 CA95008-100 CA95029-586
Proteose Peptone A, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Proteose Peptone A is recommended for cultivation of fastidious pathogens and particularly for bulk production of antibiotics, enzymes, veterinary preparations and bacterial toxins especially diphtheria toxin.