VWR® Centrifuge Cart
Supplier: VWR
VWR®
90697
75994-409EA 4524.25 CAD
75994-409
VWR® Centrifuge Cart
Trolleys
Centrifuge cart is made of high density polyethylene and ACP.
  • Weight capacity of 500 pounds on top
  • One drawer and four total locking casters
  • Latch handle safely secures drawer
  • Full extension drawer
Centrifuge cart stabilizes equipment while still allowing for mobility. The four total locking 4" casters gives you the ability to lock down your centrifuge while in use without losing mobility. Constructed from corrosive resistant HDPE, this cart has a total weight capacity of 500lbs.