Nutrient Agar 1.5%, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
M087-500G M087-500G M087-2.5KG M087-5KG
CA95020-228EA 1027.58 CAD
CA95020-228 95020-228 CA89222-502 CA89222-504
Nutrient Agar 1.5%, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Nutrient Agar is recommended for cultivation of bacteria not requiring a highly nutritious medium can be enriched with blood, ascitic fluid or other enriching fluids.
  • Final pH ( at 25 °C) is 7.3 ± 0.2
  • Cream to yellow homogeneous free flowing powder