Bile, Dried, Purified, HiMedia
Supplier: HiMedia Laboratories
RM010-10KG RM010-2.5KG RM010-500G RM010-2.5KG RM010-25KG RM010-5KG RM010-10KG RM010-5KG RM010-25KG RM010-500G
CA89229-584EA 38597.47 CAD
CA89229-584 89229-584 89229-586 CA61001-534 CA89229-590 89229-590 61001-534 CA89229-588 89229-588 CA89229-586
Bile, Dried, Purified, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Bile is obtained from purified fresh bile dessicated under controlled conditions of temperature that helps to ensure a uniform and reproducible product
  • Greenish yellow, homogeneous, free flowing powder having bile like odour
  • Final pH: 6.50 - 7.50
A selectively inhibitory agent in microbiological culture media.