Dual Click-Tite Containers, Therapak®
Supplier: Therapak, LLC
740095 740004 740005
76015-024CS 670.19 CAD
76015-024 76015-040 76015-042
Dual Click-Tite Containers, Therapak®
Sample Containers General Purpose Containers
Therapak’s Dual Click-Tite containers are ideal for maintaining the integrity of biological specimens during transport and storage.
  • Polypropylene containers are 95 kPa tested
  • Leak-proof Click-Tite cap seal design
  • Available with temperature indicator strips
Dual Click-Tite caps 'click' twice when closed, indicating the container has been closed securely and is leak-resistant for transport. Containers are stackable, graduated and include top and bottom knurls for ease of gripping with gloves. Sterile containers are complete with tamper evident label.