Delong Shaker Flasks without Baffles, Chemglass
Supplier: Chemglass
CLS-2038-01 CLS-2038-02 CLS-2038-03 CLS-2038-04 CLS-2038-06 CLS-2038-07 CLS-2038-08 CLS-2038-10 CLS-2038-11
30623-188EA 115.98 CAD
30623-188 30623-190 30623-192 30623-194 30623-196 30623-198 30623-200 30623-202 30623-204
Delong Shaker Flasks without Baffles, Chemglass
Flasks Erlenmeyer Flasks
Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures.
  • Graduated to approximate volumes
  • Made of Class A borosilicate glass Type 1
Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure.
Ordering information: Supplied with a stainless steel closure.