Bacillus laterosporus BbsI-HF, Restriction Enzymes, New England Biolabs

Supplier: New England Biolabs (NEB)

R3539L R3539S
CA103258-506EA 480 CAD
CA103258-506 CA103307-268
Bacillus laterosporus BbsI-HF, Restriction Enzymes, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes

An E. coli strain that carries the cloned and modified BbsI gene from Bacillus laterosporus (C. Story) is used as source.


One unit is defined as the amount of enzyme required to digest 1 µg of λ DNA in 1 hour at 37 °C in a total reaction volume of 50 µl.


Delivery information: It is supplied with CutSmart® Buffer (10X) and Gel Loading Dye, Purple (6X)

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Enzyme name BbsI-HF
Source Bacillus laterosporus
Storage conditions −20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR