Potassium Sodium Tartrate

Supplier: ALDON CORP SE

PP0755-100G PP0755-500G
470229-142EA 24.55 CAD
470229-142 470229-144
Potassium Sodium Tartrate
Chemicals for Science Education

CAS Number: 6381-59-5
Formula Weight: 282.23
Formula: KNaC4H4O6·4H2O
Hazard Info: Oxidizer, Toxic
Density (g/mL): 2.065
Boiling Point (°C): 105-130
Freezing Point (°C): 36.4
Solubility: Water and Alcohol
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR