SP Wilmad-LabGlass Straight Powder Funnels, SP Industries
Supplier: SP Industries
LG-8300-108 LG-8300-102 LG-8300-106 LG-8300-110
14202-768EA 124.3 CAD
14202-768 14202-028 14202-386 14206-388
SP Wilmad-LabGlass Straight Powder Funnels, SP Industries
Funnels Powder Funnels
These straight funnels are used for the introduction of liquids or powders into containers with the same Standard Taper joint.
The funnel design promotes free flow and provides a smooth surface for the transfer of material.